Product Details

SNP ID
rs180764804
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57493253 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACATGCTGGGAAAAAGACGTATGA[A/G]TGCAGTGAATGTGGGAAGTTATTTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF419 PubMed Links

Gene Details

Gene
ZNF419
Gene Name
zinc finger protein 419
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098491.1 900 Silent Mutation GAA,GAG E233E NP_001091961.1
NM_001098492.1 900 Silent Mutation GAA,GAG E220E NP_001091962.1
NM_001098493.1 900 Silent Mutation GAA,GAG E219E NP_001091963.1
NM_001098494.1 900 Silent Mutation GAA,GAG E200E NP_001091964.1
NM_001098495.1 900 Silent Mutation GAA,GAG E187E NP_001091965.1
NM_001098496.1 900 Silent Mutation GAA,GAG E186E NP_001091966.1
NM_001291743.1 900 Silent Mutation GAA,GAG E199E NP_001278672.1
NM_001291744.1 900 Silent Mutation GAA,GAG E144E NP_001278673.1
NM_001291745.1 900 Intron NP_001278674.1
NM_024691.3 900 Silent Mutation GAA,GAG E232E NP_078967.3
Gene
ZNF773
Gene Name
zinc finger protein 773
There are no transcripts associated with this gene.

View Full Product Details