Product Details

SNP ID
rs181384896
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:56911192 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGACCCTCCGATATGTTCCAGAGG[A/T]ATCCAAAGACAAAGTTATCTCAGAT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MIER3 PubMed Links

Gene Details

Gene
MIER3
Gene Name
MIER family member 3
There are no transcripts associated with this gene.

Gene
SETD9
Gene Name
SET domain containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171990.2 522 Missense Mutation GAA,GTA E41V NP_001165461.1
NM_001323018.1 522 Missense Mutation GAA,GTA E15V NP_001309947.1
NM_001323019.1 522 Missense Mutation GAA,GTA E41V NP_001309948.1
NM_001323020.1 522 Missense Mutation GAA,GTA E15V NP_001309949.1
NM_001323022.1 522 Intron NP_001309951.1
NM_153706.3 522 Missense Mutation GAA,GTA E41V NP_714917.2
XM_005248426.4 522 Intron XP_005248483.1
XM_011543139.2 522 Missense Mutation GAA,GTA E9V XP_011541441.1
XM_011543140.2 522 UTR 5 XP_011541442.1

View Full Product Details