Product Details
- SNP ID
-
rs181679827
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:14611014 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCATCTTTTCCTTTTTTTTTTTTT[C/T]CCAAACAAAAGCTTTGAAAGTGGTG
- Phenotype
-
MIM: 606101
MIM: 616838
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ADGRE3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs535841855] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADGRE3
- Gene Name
- adhesion G protein-coupled receptor E3
- Gene
- CLEC17A
- Gene Name
- C-type lectin domain family 17 member A
View Full Product Details