Product Details

SNP ID
rs181668842
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27278281 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCACTTAAACCACCACCTGAGCAG[A/G]ATAGTGGGAGAAAATATCAGCAGAA
Phenotype
MIM: 613946 MIM: 602878 MIM: 606474
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJC5G PubMed Links

Gene Details

Gene
DNAJC5G
Gene Name
DnaJ heat shock protein family (Hsp40) member C5 gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303127.1 825 Missense Mutation AAT,GAT N157D NP_001290056.1
NM_001303128.1 825 Silent Mutation AGA,AGG R69R NP_001290057.1
NM_173650.2 825 Missense Mutation AAT,GAT N157D NP_775921.1
XM_006711995.3 825 Missense Mutation AAT,GAT N157D XP_006712058.1
XM_017003872.1 825 Missense Mutation AAT,GAT N157D XP_016859361.1
XM_017003873.1 825 Missense Mutation AAT,GAT N157D XP_016859362.1
XM_017003874.1 825 Missense Mutation AAT,GAT N157D XP_016859363.1
Gene
SLC30A3
Gene Name
solute carrier family 30 member 3
There are no transcripts associated with this gene.

Gene
TRIM54
Gene Name
tripartite motif containing 54
There are no transcripts associated with this gene.

View Full Product Details