Product Details

SNP ID
rs181679927
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67200529 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGCGTGGTGTCCTGGGAGACTCT[C/G]AGCATCCCCTTTGTGAGGAAGGTGG
Phenotype
MIM: 600659 MIM: 606422 MIM: 613701
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
E2F4 PubMed Links

Gene Details

Gene
E2F4
Gene Name
E2F transcription factor 4
There are no transcripts associated with this gene.

Gene
ELMO3
Gene Name
engulfment and cell motility 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024712.3 694 Silent Mutation CTC,CTG L217L NP_078988.2
Gene
LOC105369155
Gene Name
uncharacterized LOC105369155
There are no transcripts associated with this gene.

Gene
LRRC29
Gene Name
leucine rich repeat containing 29
There are no transcripts associated with this gene.

Gene
MIR328
Gene Name
microRNA 328
There are no transcripts associated with this gene.

View Full Product Details