Product Details

SNP ID
rs181724464
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:75073671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCCGGCCGCCGTCCAGTGCCCTC[G/T]CCCCAGCCCCGGCCCGCTCAGCCGC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RCC1L PubMed Links

Gene Details

Gene
RCC1L
Gene Name
RCC1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281441.1 211 Silent Mutation AGA,CGA R23R NP_001268370.1
NM_030798.4 211 Silent Mutation AGA,CGA R23R NP_110425.2
NM_148842.2 211 Silent Mutation AGA,CGA R23R NP_683682.1

View Full Product Details