Product Details

SNP ID
rs181471295
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:129744053 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAATCTCTCAGCCTACATCATCCC[A/T]GGCACAGATGCTCACATGGTAAGAG
Phenotype
MIM: 300145
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
XPNPEP2 PubMed Links

Gene Details

Gene
XPNPEP2
Gene Name
X-prolyl aminopeptidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003399.5 460 Silent Mutation CCA,CCT P72P NP_003390.4

View Full Product Details