Product Details

SNP ID
rs181506797
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:120687338 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGTTGAGGGAACCGCTGTGGCGCT[C/T]CTGCGACTGCTGCTGCTGCTGCTGC
Phenotype
MIM: 605563 MIM: 613802
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CABP1 PubMed Links

Gene Details

Gene
CABP1
Gene Name
calcium binding protein 1
There are no transcripts associated with this gene.

Gene
MLEC
Gene Name
malectin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303627.1 215 Intron NP_001290556.1
NM_001303628.1 215 Silent Mutation CTC,CTT L14L NP_001290557.1
NM_014730.3 215 Silent Mutation CTC,CTT L14L NP_055545.1
XM_011539031.1 215 Intron XP_011537333.1
XM_011539032.1 215 Intron XP_011537334.1

View Full Product Details