Product Details

SNP ID
rs181255243
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:122473844 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTCTGGACTGGAGGCATGTCCAG[A/C]TTCTGATTTCTTTGTAAAAATCTCT
Phenotype
MIM: 616381
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZCCHC8 PubMed Links

Gene Details

Gene
ZCCHC8
Gene Name
zinc finger CCHC-type containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017612.4 1930 Missense Mutation GCT,TCT A593S NP_060082.2
XM_006719491.2 1930 Missense Mutation GCT,TCT A516S XP_006719554.1
XM_011538555.2 1930 Missense Mutation GCT,TCT A494S XP_011536857.1
XM_017019606.1 1930 Missense Mutation GCT,TCT A502S XP_016875095.1

View Full Product Details