Product Details

SNP ID
rs181315683
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:64001956 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGCCAATGAGTTCCTGGACGGGCC[A/G]GATCTGTGCTGTTGTGTAAGAGCTC
Phenotype
MIM: 146630
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ICAM2 PubMed Links

Gene Details

Gene
ICAM2
Gene Name
intercellular adhesion molecule 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000873.3 706 Intron NP_000864.2
NM_001099786.1 706 Intron NP_001093256.1
NM_001099787.1 706 Intron NP_001093257.1
NM_001099788.1 706 Intron NP_001093258.1
NM_001099789.1 706 Intron NP_001093259.1
Gene
PRR29
Gene Name
proline rich 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164257.1 706 UTR 3 NP_001157729.1
NM_001191029.1 706 Missense Mutation CAG,CGG Q231R NP_001177958.1
NM_001191030.1 706 UTR 3 NP_001177959.1
NM_001191031.1 706 UTR 3 NP_001177960.1
XM_005257805.4 706 UTR 3 XP_005257862.1
XM_011525468.2 706 Missense Mutation CAG,CGG Q209R XP_011523770.1
XM_011525469.1 706 UTR 3 XP_011523771.1
XM_017025315.1 706 Missense Mutation CAG,CGG Q144R XP_016880804.1
Gene
PRR29-AS1
Gene Name
PRR29 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details