Product Details

SNP ID
rs182141445
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:51830595 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGCAGGCCCTGTTGAGGGTCTAC[A/G]TCATCCAGGAGCAGGCGACGGTCAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IQCF3 PubMed Links

Gene Details

Gene
IQCF3
Gene Name
IQ motif containing F3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085479.2 1005 Missense Mutation ATC,GTC I87V NP_001078948.1
NM_001207023.1 1005 Missense Mutation ATC,GTC I87V NP_001193952.1

View Full Product Details