Product Details

SNP ID
rs182291591
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:53849670 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTCCCAGGGCCATCTCCAGAGT[A/G]GATGCTACAACATGATCTAATCCCC
Phenotype
MIM: 605458
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHDH PubMed Links

Gene Details

Gene
CHDH
Gene Name
choline dehydrogenase
There are no transcripts associated with this gene.

Gene
IL17RB
Gene Name
interleukin 17 receptor B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018725.3 137 Nonsense Mutation TAG,TGG *34W NP_061195.2
XM_005265310.4 137 Nonsense Mutation TAG,TGG *34W XP_005265367.1
XM_005265311.4 137 Nonsense Mutation TAG,TGG *34W XP_005265368.1
XM_005265312.4 137 Nonsense Mutation TAG,TGG *34W XP_005265369.1
XM_011533941.2 137 Nonsense Mutation TAG,TGG *34W XP_011532243.1
XM_017006804.1 137 UTR 5 XP_016862293.1
XM_017006805.1 137 UTR 5 XP_016862294.1
XM_017006806.1 137 UTR 5 XP_016862295.1
XM_017006807.1 137 Nonsense Mutation TAG,TGG *34W XP_016862296.1

View Full Product Details