Product Details
- SNP ID
-
rs182263854
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:49625202 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATCCACCTATATTTCTCAAACGAA[A/G]TGAAGAAAATAGTTCAAAATTTGTG
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DTWD1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11539519] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DTWD1
- Gene Name
- DTW domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001144955.1 |
231 |
Missense Mutation |
AAT,AGT |
N12S |
NP_001138427.1 |
NM_020234.5 |
231 |
Missense Mutation |
AAT,AGT |
N12S |
NP_064619.2 |
XM_011521815.1 |
231 |
Missense Mutation |
AAT,AGT |
N12S |
XP_011520117.1 |
XM_017022419.1 |
231 |
Missense Mutation |
AAT,AGT |
N12S |
XP_016877908.1 |
XM_017022420.1 |
231 |
Missense Mutation |
AAT,AGT |
N12S |
XP_016877909.1 |
XM_017022421.1 |
231 |
Missense Mutation |
AAT,AGT |
N12S |
XP_016877910.1 |
XM_017022422.1 |
231 |
Missense Mutation |
AAT,AGT |
N12S |
XP_016877911.1 |
XM_017022423.1 |
231 |
Missense Mutation |
AAT,AGT |
N12S |
XP_016877912.1 |
XM_017022424.1 |
231 |
Missense Mutation |
AAT,AGT |
N12S |
XP_016877913.1 |
XM_017022425.1 |
231 |
Missense Mutation |
AAT,AGT |
N12S |
XP_016877914.1 |
XM_017022426.1 |
231 |
Missense Mutation |
AAT,AGT |
N12S |
XP_016877915.1 |
XM_017022427.1 |
231 |
UTR 5 |
|
|
XP_016877916.1 |
XM_017022428.1 |
231 |
UTR 5 |
|
|
XP_016877917.1 |
XM_017022429.1 |
231 |
UTR 5 |
|
|
XP_016877918.1 |
XM_017022430.1 |
231 |
UTR 5 |
|
|
XP_016877919.1 |
XM_017022431.1 |
231 |
Intron |
|
|
XP_016877920.1 |
- Gene
- FAM227B
- Gene Name
- family with sequence similarity 227 member B
There are no transcripts associated with this gene.
View Full Product Details