Product Details

SNP ID
rs182259769
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:110386331 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCCCCAAGAATTACTTACCTTT[A/T]ATCAGATAAGGATCCAACATCTGTG
Phenotype
MIM: 606949
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
ANAPC7 PubMed Links

Gene Details

Gene
ANAPC7
Gene Name
anaphase promoting complex subunit 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001137664.1 916 Silent Mutation ATA,ATT I305I NP_001131136.1
NM_016238.2 916 Silent Mutation ATA,ATT I305I NP_057322.2
XM_005253903.3 916 Silent Mutation ATA,ATT I84I XP_005253960.1
XM_011538454.2 916 Silent Mutation ATA,ATT I285I XP_011536756.1
XM_017019425.1 916 Silent Mutation ATA,ATT I305I XP_016874914.1
XM_017019426.1 916 Silent Mutation ATA,ATT I204I XP_016874915.1
XM_017019427.1 916 Silent Mutation ATA,ATT I204I XP_016874916.1
XM_017019428.1 916 Silent Mutation ATA,ATT I84I XP_016874917.1

View Full Product Details