Product Details

SNP ID
rs182264437
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:144379913 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGCCCACAGGCTGGCTGGAGGGC[A/G]TGAGGCTCTCAGACGGGGAGCGAGG
Phenotype
MIM: 600888
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF5 PubMed Links

Gene Details

Gene
ARHGEF5
Gene Name
Rho guanine nucleotide exchange factor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005435.3 4769 Missense Mutation ATG,GTG M1551V NP_005426.2
XM_017012623.1 4769 Intron XP_016868112.1

View Full Product Details