Product Details

SNP ID
rs181966984
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:90461326 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGGTATTGAAGTGCCCAGGAAG[A/C]ACCTTCTCTACCAGACCTCTGTCCA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MBLAC2 PubMed Links

Gene Details

Gene
MBLAC2
Gene Name
metallo-beta-lactamase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203406.1 1157 Silent Mutation GTG,GTT V227V NP_981951.1

View Full Product Details