Product Details

SNP ID
rs182093138
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:13144512 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGAACGAGAAGGTCAGGGGTCA[A/C]GGGGAGAGGGTACCACCTACAACCT
Phenotype
MIM: 610672 MIM: 603765
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IER2 PubMed Links

Gene Details

Gene
IER2
Gene Name
immediate early response 2
There are no transcripts associated with this gene.

Gene
NACC1
Gene Name
nucleus accumbens associated 1
There are no transcripts associated with this gene.

Gene
STX10
Gene Name
syntaxin 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271609.1 939 Intron NP_001258538.1
NM_001271610.1 939 Silent Mutation CCA,CCC P246P NP_001258539.1
NM_001271611.1 939 Intron NP_001258540.1
NM_003765.2 939 Intron NP_003756.1
XM_011528397.2 939 Intron XP_011526699.1

View Full Product Details