Product Details
- SNP ID
-
rs181802264
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:49208400 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACAGCCGAGTCGGTTTCGTGAAGGA[A/G]ATTCCACATCTCGGCGCGAGCAGAG
- Phenotype
-
MIM: 600934
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FOLH1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs200069827] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FOLH1
- Gene Name
- folate hydrolase (prostate-specific membrane antigen) 1
View Full Product Details