Product Details

SNP ID
rs181841908
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:17373225 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACATCGCCCAGCCCTTCCGTGGAT[A/G]CGGATAAGGTAAGCCTCAGGGGAAG
Phenotype
MIM: 605347 MIM: 610363
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PADI4 PubMed Links

Gene Details

Gene
PADI4
Gene Name
peptidyl arginine deiminase 4
There are no transcripts associated with this gene.

Gene
PADI6
Gene Name
peptidyl arginine deiminase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207421.4 336 Missense Mutation ACG,GCG T96A NP_997304.3

View Full Product Details