Product Details

SNP ID
rs182794327
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:109250587 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCACCAGAAGAGTCCCTGGGTGGG[C/T]GTCGGAAAAGGAATGTAAATACAGC
Phenotype
MIM: 604265
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CELSR2 PubMed Links

Gene Details

Gene
CELSR2
Gene Name
cadherin EGF LAG seven-pass G-type receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001408.2 569 Missense Mutation CGT,TGT R170C NP_001399.1
XM_005270580.4 569 Missense Mutation CGT,TGT R170C XP_005270637.1

View Full Product Details