Product Details

SNP ID
rs182799009
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:159713585 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGCACTTCATACTTCAGTGCCCG[C/T]CAGTTCAGGACATTAGGACTGAAGG
Phenotype
MIM: 123260
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRP PubMed Links

Gene Details

Gene
CRP
Gene Name
C-reactive protein, pentraxin-related
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000567.2 719 Nonsense Mutation TGA,TGG *205W NP_000558.2
XM_011509207.2 719 Nonsense Mutation TGA,TGG *205W XP_011507509.1

View Full Product Details