Product Details

SNP ID
rs182522714
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136496082 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGGGCGCGCCGTTTACTTGAAGG[A/C]CTCCGGAATGCGGGCGATCTGGGAC
Phenotype
MIM: 190198
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NOTCH1 PubMed Links

Gene Details

Gene
NOTCH1
Gene Name
notch 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017617.4 6949 Missense Mutation GCC,TCC A2553S NP_060087.3
XM_011518717.2 6949 Missense Mutation GCC,TCC A2312S XP_011517019.2

View Full Product Details