Product Details

SNP ID
rs182380930
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:66963300 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGGGCATTCCATTTGCCCAGCC[A/G]CCACTGGGCCCTGACCGGTTCTCAG
Phenotype
MIM: 605279
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CES3 PubMed Links

Gene Details

Gene
CES3
Gene Name
carboxylesterase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185176.1 283 Intron NP_001172105.1
NM_001185177.1 283 Silent Mutation CCA,CCG P68P NP_001172106.1
NM_024922.5 283 Silent Mutation CCA,CCG P68P NP_079198.2
XM_011522975.2 283 Intron XP_011521277.1
XM_011522976.2 283 Silent Mutation CCA,CCG P68P XP_011521278.1

View Full Product Details