Product Details

SNP ID
rs183023945
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:77983405 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAATTCTGATTACCAAGGCAAAAT[G/T]GGGTAATGCTATACAATGGGGGCAG
Phenotype
MIM: 610741
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TNRC6C PubMed Links

Gene Details

Gene
TNRC6C
Gene Name
trinucleotide repeat containing 6C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142640.1 Intron NP_001136112.1
NM_018996.3 Intron NP_061869.2
XM_006721996.3 Intron XP_006722059.3
XM_006721997.3 Intron XP_006722060.3
XM_006722000.3 Intron XP_006722063.3
XM_011525088.2 Intron XP_011523390.1
XM_017024906.1 Intron XP_016880395.1
XM_017024907.1 Intron XP_016880396.1
XM_017024908.1 Intron XP_016880397.1

View Full Product Details