Product Details

SNP ID
rs183232880
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:10607286 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGTGGGCCTGTCCTCAGGGACT[C/G]GGCTGCTGCTTTCAGAAGCCTCCTC
Phenotype
MIM: 608581
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RP1L1 PubMed Links

Gene Details

Gene
RP1L1
Gene Name
retinitis pigmentosa 1-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178857.5 7041 Missense Mutation CCA,CGA P2271R NP_849188.4

View Full Product Details