Product Details

SNP ID
rs183241610
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:27462363 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGCGCAGCAGGGAGCTCTCGGG[A/G]ATGCCAGGCCTGGTCATGATGTGGT
Phenotype
MIM: 603246
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GTF3C1 PubMed Links

Gene Details

Gene
GTF3C1
Gene Name
general transcription factor IIIC subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286242.1 5895 Silent Mutation ATC,ATT I1991I NP_001273171.1
NM_001520.3 5895 Silent Mutation ATC,ATT I2016I NP_001511.2
XM_017023188.1 5895 Silent Mutation ATC,ATT I1953I XP_016878677.1
Gene
IL21R-AS1
Gene Name
IL21R antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details