Product Details

SNP ID
rs183289121
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:130205322 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTCTCTATTATAACTTGGCAAGT[A/G]TGAGTTTTACGTTGACTTAAGTGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF280C PubMed Links

Gene Details

Gene
ZNF280C
Gene Name
zinc finger protein 280C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017666.4 2339 Silent Mutation CAC,CAT H712H NP_060136.1
XM_006724765.3 2339 Silent Mutation CAC,CAT H663H XP_006724828.1

View Full Product Details