Product Details

SNP ID
rs183275634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:38866432 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGGGCTCAGGGGCCGTGGGAGGT[A/G]CCGGGCCGGCAGCAGCTGTGACCTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CBX6 PubMed Links

Gene Details

Gene
CBX6
Gene Name
chromobox 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303494.1 1160 Missense Mutation GCA,GTA A321V NP_001290423.1
NM_014292.4 1160 Missense Mutation GCA,GTA A339V NP_055107.3

View Full Product Details