Product Details

SNP ID
rs183385711
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:28681983 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCATTTTCTTTGTACTCCTTTAG[A/G]ACAAGGAAACAGGTATCAGCATGAT
Phenotype
MIM: 604444
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BAMBI PubMed Links

Gene Details

Gene
BAMBI
Gene Name
BMP and activin membrane bound inhibitor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012342.2 768 Missense Mutation GAA,GGA E122G NP_036474.1

View Full Product Details