Product Details

SNP ID
rs183376032
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:149820664 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATGCAACCATGAGAAGCAGCAGA[A/G]CAGTGATAGCCAAACCCAGCGCCAG
Phenotype
MIM: 176851
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRP11 PubMed Links

Gene Details

Gene
LRP11
Gene Name
LDL receptor related protein 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032832.5 1712 Missense Mutation GCT,GTT A463V NP_116221.3
XM_011536194.2 1712 Missense Mutation GCT,GTT A431V XP_011534496.1
XM_017011380.1 1712 Intron XP_016866869.1
Gene
PCMT1
Gene Name
protein-L-isoaspartate (D-aspartate) O-methyltransferase
There are no transcripts associated with this gene.

View Full Product Details