Product Details

SNP ID
rs183104887
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:96024592 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGTGAGTGCTGCAGCAGGCTCC[A/G]CAGCTGCCCAGAGAAGCCTACATCA
Phenotype
MIM: 616431
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAHD2CP PubMed Links

Gene Details

Gene
FAHD2CP
Gene Name
fumarylacetoacetate hydrolase domain containing 2C, pseudogene
There are no transcripts associated with this gene.

Gene
GPAT2
Gene Name
glycerol-3-phosphate acyltransferase 2, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321525.1 Intron NP_001308454.1
NM_001321526.1 Intron NP_001308455.1
NM_001321527.1 Intron NP_001308456.1
NM_001321528.1 Intron NP_001308457.1
NM_001321529.1 Intron NP_001308458.1
NM_001321530.1 Intron NP_001308459.1
NM_001321531.1 Intron NP_001308460.1
NM_207328.3 Intron NP_997211.2
XM_005263888.3 Intron XP_005263945.1
XM_006712308.3 Intron XP_006712371.1
XM_006712311.3 Intron XP_006712374.1
XM_006712312.3 Intron XP_006712375.1
XM_006712313.3 Intron XP_006712376.1
XM_017003424.1 Intron XP_016858913.1
XM_017003425.1 Intron XP_016858914.1
XM_017003426.1 Intron XP_016858915.1
Gene
LOC107984110
Gene Name
fumarylacetoacetate hydrolase domain-containing protein 2B-like
There are no transcripts associated with this gene.

View Full Product Details