Product Details

SNP ID
rs183168709
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:58428446 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCTGTTGGAACGGGGACTAGTT[C/T]GAATGGAACATCACGTGGTATGTGA
Phenotype
MIM: 610178 MIM: 607384
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIAA0586 PubMed Links

Gene Details

Gene
KIAA0586
Gene Name
KIAA0586
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244189.1 428 Missense Mutation TCG,TTG S73L NP_001231118.1
NM_001244190.1 428 Missense Mutation TCG,TTG S46L NP_001231119.1
NM_001244191.1 428 Intron NP_001231120.1
NM_001244192.1 428 Intron NP_001231121.1
NM_001244193.1 428 Intron NP_001231122.1
Gene
TIMM9
Gene Name
translocase of inner mitochondrial membrane 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304485.1 428 Intron NP_001291414.1
NM_001304486.1 428 Intron NP_001291415.1
NM_001304487.1 428 Intron NP_001291416.1
NM_001304488.1 428 Intron NP_001291417.1
NM_001304489.1 428 Intron NP_001291418.1
NM_001304490.1 428 Intron NP_001291419.1
NM_001304491.1 428 Intron NP_001291420.1
NM_012460.3 428 Intron NP_036592.1

View Full Product Details