Product Details
- SNP ID
-
rs183223952
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:127549513 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTCAGTAAAACTCAATCTCATCCT[A/G]GTTCTGTCGCTGTCCACAATGCATG
- Phenotype
-
MIM: 609513
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NPS
PubMed Links
Gene Details
- Gene
- NPS
- Gene Name
- neuropeptide S
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001030013.1 |
53 |
Silent Mutation |
CTA,CTG |
L11L |
NP_001025184.1 |
View Full Product Details