Product Details

SNP ID
rs183779548
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144012620 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGCGGCTCATTCGGGAGAATCA[C/T]GAGCTCAAGTCAGCGATCAAGACTC
Phenotype
MIM: 610874
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPATC1 PubMed Links

Gene Details

Gene
SPATC1
Gene Name
spermatogenesis and centriole associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134374.1 207 Silent Mutation CAC,CAT H35H NP_001127846.1
NM_198572.2 207 Silent Mutation CAC,CAT H35H NP_940974.2
XM_011517021.1 207 Silent Mutation CAC,CAT H35H XP_011515323.1
XM_011517022.2 207 Intron XP_011515324.1
XM_011517023.2 207 Intron XP_011515325.1
XM_011517024.2 207 Intron XP_011515326.1
XM_011517025.2 207 Intron XP_011515327.1
XM_017013399.1 207 Intron XP_016868888.1

View Full Product Details