Product Details
- SNP ID
-
rs183837737
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:151282385 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAATTCCACCCTTTTGTTGGGTGAC[C/T]GGATGGGGAAAAGTTAAGGAAAGTT
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PRSS48
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs17027509] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PRSS48
- Gene Name
- protease, serine 48
View Full Product Details