Product Details

SNP ID
rs183788679
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:103403240 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTCACTGAAATCTGTGATGCCTA[C/T]ACCAAAAAGCTGAATGAGCAGGTGA
Phenotype
MIM: 612333 MIM: 615204
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR1307 PubMed Links

Gene Details

Gene
MIR1307
Gene Name
microRNA 1307
There are no transcripts associated with this gene.

Gene
PDCD11
Gene Name
programmed cell death 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014976.1 484 Silent Mutation TAC,TAT Y119Y NP_055791.1
XM_005269647.3 484 Silent Mutation TAC,TAT Y119Y XP_005269704.1
XM_011539538.2 484 Silent Mutation TAC,TAT Y119Y XP_011537840.1
XM_011539539.2 484 Silent Mutation TAC,TAT Y119Y XP_011537841.1
XM_011539540.1 484 Silent Mutation TAC,TAT Y36Y XP_011537842.1
Gene
USMG5
Gene Name
up-regulated during skeletal muscle growth 5 homolog (mouse)
There are no transcripts associated with this gene.

View Full Product Details