Product Details
- SNP ID
-
rs183592454
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:57230913 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACAGGGGCTGGACAGGCCAGGAGAG[C/G]CTGTCGGACAGTGATCCTGAGATGT
- Phenotype
-
MIM: 604637
MIM: 138450
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NDUFA4L2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs73338162] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NDUFA4L2
- Gene Name
- NDUFA4, mitochondrial complex associated like 2
There are no transcripts associated with this gene.
- Gene
- NXPH4
- Gene Name
- neurexophilin 4
There are no transcripts associated with this gene.
- Gene
- SHMT2
- Gene Name
- serine hydroxymethyltransferase 2
View Full Product Details