Product Details

SNP ID
rs183598373
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:12337430 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTATTCGTACTTCATCATCTATCA[A/G]TCTTGCAGTGGCTTCACTGTAAGGT
Phenotype
MIM: 604581 MIM: 615103
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AFG3L2 PubMed Links

Gene Details

Gene
AFG3L2
Gene Name
AFG3 like matrix AAA peptidase subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006796.2 2280 Silent Mutation CTG,TTG L696L NP_006787.2
XM_011525601.2 2280 Silent Mutation CTG,TTG L629L XP_011523903.1
Gene
TUBB6
Gene Name
tubulin beta 6 class V
There are no transcripts associated with this gene.

View Full Product Details