Product Details

SNP ID
rs183621176
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:82732854 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGTCCCCGCAGTACGGAGGCTCC[A/G]GCGGGGAACACGTCGAGAGGCTCGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM181B PubMed Links

Gene Details

Gene
FAM181B
Gene Name
family with sequence similarity 181 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175885.3 1011 Silent Mutation GCC,GCT A292A NP_787081.2

View Full Product Details