Product Details

SNP ID
rs184061405
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:196779898 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGTTGCCTGCCATCCTGGCTAC[A/G]GTCTTCCAAAAGCGCAGACCACAGT
Phenotype
MIM: 605336
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CFHR3 PubMed Links

Gene Details

Gene
CFHR3
Gene Name
complement factor H related 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166624.1 442 Missense Mutation AGT,GGT S119G NP_001160096.1
NM_021023.5 442 Missense Mutation AGT,GGT S119G NP_066303.2

View Full Product Details