Product Details
- SNP ID
-
rs184374125
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:50419077 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGAGGTGCGGCGGGCCCCGCCTGC[A/T]GCGGCGCAGGCAGCCCAGGCCCCCC
- Phenotype
-
MIM: 614804
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
DMRTA2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3862270] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DMRTA2
- Gene Name
- DMRT like family A2
View Full Product Details