Product Details

SNP ID
rs184360961
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:15109497 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCTTCCCCCCATCGCATGTGCCC[C/T]TCCCTCTGAATGGTATCTTCCTGTT
Phenotype
MIM: 612664
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RERG PubMed Links

Gene Details

Gene
RERG
Gene Name
RAS like estrogen regulated growth inhibitor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190726.1 553 Silent Mutation GAA,GAG E52E NP_001177655.1
NM_032918.2 553 Silent Mutation GAA,GAG E71E NP_116307.1
XM_017020121.1 553 Silent Mutation GAA,GAG E68E XP_016875610.1

View Full Product Details