Product Details
- SNP ID
-
rs184182963
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:3815406 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAAGGAGCCCGAGCGCCGCGTCAG[C/T]GCTTGGCGTACGTCCTGCTGGAGCT
- Phenotype
-
MIM: 616690
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CEP104
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs12728401] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CEP104
- Gene Name
- centrosomal protein 104
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014704.3 |
3235 |
Missense Mutation |
CAC,CGC |
H925R |
NP_055519.1 |
XM_005244815.4 |
3235 |
Missense Mutation |
CAC,CGC |
H961R |
XP_005244872.1 |
XM_011542473.2 |
3235 |
Missense Mutation |
CAC,CGC |
H967R |
XP_011540775.1 |
XM_011542474.2 |
3235 |
Missense Mutation |
CAC,CGC |
H931R |
XP_011540776.1 |
XM_011542475.2 |
3235 |
Missense Mutation |
CAC,CGC |
H909R |
XP_011540777.1 |
XM_011542476.2 |
3235 |
Missense Mutation |
CAC,CGC |
H901R |
XP_011540778.1 |
XM_011542477.2 |
3235 |
Missense Mutation |
CAC,CGC |
H843R |
XP_011540779.1 |
XM_011542478.1 |
3235 |
Intron |
|
|
XP_011540780.1 |
XM_017002917.1 |
3235 |
Missense Mutation |
CAC,CGC |
H903R |
XP_016858406.1 |
XM_017002918.1 |
3235 |
Missense Mutation |
CAC,CGC |
H867R |
XP_016858407.1 |
XM_017002919.1 |
3235 |
Missense Mutation |
CAC,CGC |
H859R |
XP_016858408.1 |
View Full Product Details