Product Details

SNP ID
rs184300283
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:127878182 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATTCCGTCCCTTGGAGCATATTC[C/T]ATTGGCTTTCTCTGCTCCTGCTTCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLRN3 PubMed Links

Gene Details

Gene
CLRN3
Gene Name
clarin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152311.3 630 Missense Mutation ATA,ATG I216M NP_689524.1
XM_011539274.2 630 Missense Mutation ATA,ATG I156M XP_011537576.1

View Full Product Details