Product Details

SNP ID
rs184923665
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8393377 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACACATGGCCACAGCTCAGCGTCC[A/G]GCTGGCGCCCTCCAGGTCCCGGAAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNF222 PubMed Links

Gene Details

Gene
RNF222
Gene Name
ring finger protein 222
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146684.2 479 Missense Mutation CGG,TGG R29W NP_001140156.1
XM_011523978.2 479 Missense Mutation CGG,TGG R29W XP_011522280.1
XM_011523979.2 479 Missense Mutation CGG,TGG R29W XP_011522281.1
XM_011523980.2 479 Missense Mutation CGG,TGG R29W XP_011522282.1
XM_011523981.2 479 Missense Mutation CGG,TGG R29W XP_011522283.1

View Full Product Details