Product Details

SNP ID
rs184886290
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:4746963 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCTCTGACAGTGAGGAGGAGGA[A/G]GAGGAAGAGATGGCAGCTCTGGGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C16orf71 PubMed Links

Gene Details

Gene
C16orf71
Gene Name
chromosome 16 open reading frame 71
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139170.2 1409 Silent Mutation GAA,GAG E406E NP_631909.2
XM_005255144.3 1409 Silent Mutation GAA,GAG E268E XP_005255201.1
XM_017022975.1 1409 Silent Mutation GAA,GAG E429E XP_016878464.1
XM_017022976.1 1409 Silent Mutation GAA,GAG E406E XP_016878465.1
Gene
ZNF500
Gene Name
zinc finger protein 500
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303450.1 1409 Intron NP_001290379.1
NM_021646.3 1409 Intron NP_067678.1
XM_005255243.3 1409 Intron XP_005255300.1
XM_011522453.2 1409 Intron XP_011520755.1
XM_011522454.2 1409 Intron XP_011520756.1
XM_017023121.1 1409 Intron XP_016878610.1

View Full Product Details