Product Details
- SNP ID
-
rs185056000
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:170234150 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGGCTCTGAAATGAGAGAAATTGC[A/C]GAAATTCTCCCTGCTCAGGAAAAGG
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
C5orf58
PubMed Links
Gene Details
- Gene
- C5orf58
- Gene Name
- chromosome 5 open reading frame 58
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001102609.2 |
127 |
Silent Mutation |
GCA,GCC |
A5A |
NP_001096079.1 |
NM_001305393.1 |
127 |
Silent Mutation |
GCA,GCC |
A5A |
NP_001292322.1 |
NM_001305394.1 |
127 |
Silent Mutation |
GCA,GCC |
A5A |
NP_001292323.1 |
NM_001305395.1 |
127 |
Silent Mutation |
GCA,GCC |
A5A |
NP_001292324.1 |
NM_001305396.1 |
127 |
Silent Mutation |
GCA,GCC |
A5A |
NP_001292325.1 |
NM_001305397.1 |
127 |
Silent Mutation |
GCA,GCC |
A5A |
NP_001292326.1 |
XM_011534432.2 |
127 |
Silent Mutation |
GCA,GCC |
A44A |
XP_011532734.1 |
XM_017009029.1 |
127 |
Silent Mutation |
GCA,GCC |
A44A |
XP_016864518.1 |
XM_017009030.1 |
127 |
Silent Mutation |
GCA,GCC |
A44A |
XP_016864519.1 |
XM_017009031.1 |
127 |
Silent Mutation |
GCA,GCC |
A5A |
XP_016864520.1 |
XM_017009032.1 |
127 |
Silent Mutation |
GCA,GCC |
A5A |
XP_016864521.1 |
XM_017009033.1 |
127 |
Silent Mutation |
GCA,GCC |
A5A |
XP_016864522.1 |
View Full Product Details