Product Details

SNP ID
rs185071029
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:35802485 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATCAAGAAGAGGAAGATAGTCCA[A/G]GCTCCTCAGAGGATACATCAGACCC
Phenotype
MIM: 300762
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAGEB16 PubMed Links

Gene Details

Gene
MAGEB16
Gene Name
MAGE family member B16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099921.1 457 Missense Mutation AGC,GGC S97G NP_001093391.1
XM_011545453.2 457 Missense Mutation AGC,GGC S97G XP_011543755.1
XM_011545454.2 457 Missense Mutation AGC,GGC S97G XP_011543756.1

View Full Product Details