Product Details

SNP ID
rs185036882
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:56964514 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAATCCTGCCATTCGTTCTTTTC[A/C]ACCCATTGGAATCTTCCAATGAAAT
Phenotype
MIM: 614919
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
NOA1 PubMed Links

Gene Details

Gene
NOA1
Gene Name
nitric oxide associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032313.3 1852 Missense Mutation NP_115689.1

View Full Product Details