Product Details

SNP ID
rs185005508
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:50376028 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTCACTCCTTTCTACTTACTTCA[A/G]TGGGAATACAGGAAGCTGCACCCAT
Phenotype
MIM: 300837
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DGKK PubMed Links

Gene Details

Gene
DGKK
Gene Name
diacylglycerol kinase kappa
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013742.3 3582 Missense Mutation ACT,ATT T1137I NP_001013764.1
XM_017029268.1 3582 Missense Mutation ACT,ATT T1137I XP_016884757.1

View Full Product Details